missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil, Heavy (USP/FCC), Fisher Chemical™

Catalog No. p-168739
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O1224 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O1224 Supplier Thermo Fisher Scientific Supplier No. O122 4
Only null left

Saybolt viscosity: 162 minimum

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number MFCD00131611
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Acidity Pass Test (USP)
Absorbance Pass Test (FCC)
Color White
Density 0.83g/mL
Boiling Point 260°C
Flash Point 160°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 0.845 to 0.905 (FCC/USP)
Residual Solvents Meets Requirements (USP)
Viscosity 3 CS min. (FCC), 34.5 to 150.0 mm2.S-1 (USP)
Vapor Pressure <0.1 mmHg
Grade FCC/USP
Physical Form Liquid
Chemical Name or Material Mineral Oil, Heavy
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:0
Flammability:1
Instability:0