missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Stopper for Vacuum Filtration

Catalog No. XX1014708
Change view
Click to view available options
:
Stopper No. 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
XX1014708 Stopper No. 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XX1014708 Supplier MilliporeSigma™ Supplier No. XX1014708
Only null left

Stopper No. 8 is designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration.

Millipore Glass Vacuum Filtration Accessory Product:

  • Stopper, No. 8, perforated, parylene coated, 5 Pk.
  • These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
  • This item is an accessory part for the Millipore 47 mm Classic Glass Filter Holder Kits.

Specifications

Color Clear
For Use With (Equipment) Millipore Classic Glass Filter Holder Kit
Material Silicone
For Use With (Application) Vacuum filtration
Quantity 5 Pk.
Diameter (Metric)Top 41.2 mm