missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma Millipore Stopper for Vacuum Filtration

Catalog No. XX2014718
Change view
Click to view available options
Diameter (English):
1.61 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
XX2014718 1.61 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XX2014718 Supplier MilliporeSigma Supplier No. XX2014718*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Silicone stopper with parylene coating.

These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.

Specifications

Disposable Reusable
Material Silicone
Sterility Non-sterile
No. per Pack 5/Pk.
Type Stopper
Diameter (English) 1.61 in.
Diameter (English) Inside 0.37 in.