missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma Millicell Hanging Cell Culture Inserts

Millicell™ hanging cell culture inserts with track-etched PET membrane promote natural growth and differentiation of various cell types

Supplier:  MilliporeSigma PTSP12H48

 View more versions of this product

Catalog No. PTSP12H48


Only null left

Description

Description

  • Individually blister-packaged inserts fit in most culture plates
  • TC-treated and gamma sterilized
  • Pore sizes range from 0.4 to 8.0 μm
  • Low pore density membranes are optically transparent
  • High pore density membranes are optically translucent
  • Applications include co-culture, migration and invasion assays, toxicity testing, transport and permeability assays and organotypic Air-Liquid Interface (ALI) culture
Specifications

Specifications

1.1 cm2
0.8 mL
2 x 106 pores/cm2
Cell Culture Insert
12 mm
15.5 mm
Translucent
Sterile
Individually Blister Packaged
PET
0.4 mL
3 μm
Culture Plate
15 mm
48/Pk.
TC-treated
12
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions