missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma Millicell Hanging Cell Culture Inserts

Millicell™ hanging cell culture inserts with track-etched PET membrane promote natural growth and differentiation of various cell types

Supplier:  MilliporeSigma PCHT06H48

 View more versions of this product

Catalog No. PCHT06H48


Only null left

Description

Description

  • Individually blister-packaged inserts fit in most culture plates
  • TC-treated and gamma sterilized
  • Pore sizes range from 0.4 to 8.0 μm
  • Low pore density membranes are optically transparent
  • High pore density membranes are optically translucent
  • Applications include co-culture, migration and invasion assays, toxicity testing, transport and permeability assays and organotypic Air-Liquid Interface (ALI) culture
Specifications

Specifications

4.5 cm2
4 mL
4 x 106 pores/cm2
Cell Culture Insert
24 mm
15.5 mm
Transparent
Sterile
Individually Blister Packaged
PET
2 mL
0.4 μm
Culture Plate
27 mm
48/Pk.
TC-treated
6
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions