missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma Millicell Hanging Cell Culture Inserts

Catalog No. PTSP24H48
Change view
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
5 μm
8 μm
No. of Wells:
12
24
6
Pore Density:
1 x 108 pores/cm2
2 x 105 pores/cm2
2 x 106 pores/cm2
4 x 106 pores/cm2
Color:
Translucent
Transparent
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size No. of Wells Pore Density Color
PTSP24H48 3 μm 24 2 x 106 pores/cm2 Translucent
PCEP06H48 8 μm 6 - Transparent
PCEP12H48 8 μm 12 - Transparent
PCEP24H48 8 μm 24 - Transparent
PCHT06H48 0.4 μm 6 4 x 106 pores/cm2 Transparent
PCHT12H48 0.4 μm 12 4 x 106 pores/cm2 Transparent
PCHT24H48 0.4 μm 24 4 x 106 pores/cm2 Transparent
PCSP06H48 3 μm 6 - Transparent
PCSP12H48 3 μm 12 - Transparent
PCSP24H48 3 μm 24 - Transparent
PLRP06H48 1 μm 6 - Translucent
PLRP12H48 1 μm 12 - Translucent
PLRP24H48 1 μm 24 - Translucent
PTEP06H48 8 μm 6 2 x 105 pores/cm2 Translucent
PTEP12H48 8 μm 12 2 x 105 pores/cm2 Translucent
PTHT06H48 0.4 μm 6 1 x 108 pores/cm2 Translucent
PTHT12H48 0.4 μm 12 1 x 108 pores/cm2 Translucent
PTSP06H48 3 μm 6 2 x 106 pores/cm2 Translucent
PTSP12H48 3 μm 12 2 x 106 pores/cm2 Translucent
PTRP06H48 1 μm 6 2 x 106 pores/cm2 Transparent
PTRP12H48 1 μm 12 2 x 106 pores/cm2 Transparent
PTRP24H48 1 μm 24 2 x 106 pores/cm2 Transparent
PTMP06H48 5 μm 6 - Transparent
PTMP12H48 5 μm 12 - Transparent
PTMP24H48 5 μm 24 - Transparent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
Catalog No. PTSP24H48 Supplier MilliporeSigma Supplier No. PTSP24H48

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Millicell™ hanging cell culture inserts with track-etched PET membrane promote natural growth and differentiation of various cell types

  • Individually blister-packaged inserts fit in most culture plates
  • TC-treated and gamma sterilized
  • Pore sizes range from 0.4 to 8.0 μm
  • Low pore density membranes are optically transparent
  • High pore density membranes are optically translucent
  • Applications include co-culture, migration and invasion assays, toxicity testing, transport and permeability assays and organotypic Air-Liquid Interface (ALI) culture

Specifications

Culture Area 0.3 cm2
Material PET
Volume (Metric) Well 0.4 mL
Volume (Metric) Working 0.2 mL
Pore Density 2 x 106 pores/cm2
Pore Size 3 μm
Type Cell Culture Insert
For Use With (Equipment) Culture Plate
Diameter (Metric) Inner 6.5 mm
Diameter (Metric) Outer 9 mm
Height (Metric) 15.5 mm
Quantity 48/Pk.
Color Translucent
Surface Treatment TC-treated
Sterility Sterile
No. of Wells 24
Packaging Individually Blister Packaged
Disposable Single-use
Show More Show Less