missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Millicell Hanging Cell Culture Inserts
Millicell™ hanging cell culture inserts with track-etched PET membrane promote natural growth and differentiation of various cell types
Supplier: MilliporeSigma PTMP12H48
Description
- Individually blister-packaged inserts fit in most culture plates
- TC-treated and gamma sterilized
- Pore sizes range from 0.4 to 8.0 μm
- Low pore density membranes are optically transparent
- High pore density membranes are optically translucent
- Applications include co-culture, migration and invasion assays, toxicity testing, transport and permeability assays and organotypic Air-Liquid Interface (ALI) culture
Chemical Identifiers
| PET | |
| Culture Plate | |
| 48/Pk. | |
| Sterile | |
| Single-use |
| Cell Culture Insert | |
| 15.5 mm | |
| TC-treated | |
| Individually Blister Packaged |
Specifications
| 1.1 cm2 | |
| 0.8 mL | |
| 5 μm | |
| Culture Plate | |
| 15 mm | |
| 48/Pk. | |
| TC-treated | |
| 12 | |
| Single-use |
| PET | |
| 0.4 mL | |
| Cell Culture Insert | |
| 12 mm | |
| 15.5 mm | |
| Transparent | |
| Sterile | |
| Individually Blister Packaged |