missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma Millicell Hanging Cell Culture Inserts

Catalog No. PCEP06H48
Change view
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
5 μm
8 μm
No. of Wells:
12
24
6
Color:
Translucent
Transparent
Pore Density:
1 x 108 pores/cm2
2 x 105 pores/cm2
2 x 106 pores/cm2
4 x 106 pores/cm2
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size No. of Wells Color Pore Density
PCEP06H48 8 μm 6 Transparent -
PCEP12H48 8 μm 12 Transparent -
PCEP24H48 8 μm 24 Transparent -
PCHT06H48 0.4 μm 6 Transparent 4 x 106 pores/cm2
PCHT12H48 0.4 μm 12 Transparent 4 x 106 pores/cm2
PCHT24H48 0.4 μm 24 Transparent 4 x 106 pores/cm2
PCSP06H48 3 μm 6 Transparent -
PCSP24H48 3 μm 24 Transparent -
PLRP06H48 1 μm 6 Translucent -
PLRP12H48 1 μm 12 Translucent -
PLRP24H48 1 μm 24 Translucent -
PTEP06H48 8 μm 6 Translucent 2 x 105 pores/cm2
PTEP12H48 8 μm 12 Translucent 2 x 105 pores/cm2
PTHT06H48 0.4 μm 6 Translucent 1 x 108 pores/cm2
PTHT12H48 0.4 μm 12 Translucent 1 x 108 pores/cm2
PTSP06H48 3 μm 6 Translucent 2 x 106 pores/cm2
PTSP12H48 3 μm 12 Translucent 2 x 106 pores/cm2
PTSP24H48 3 μm 24 Translucent 2 x 106 pores/cm2
PTRP06H48 1 μm 6 Transparent 2 x 106 pores/cm2
PTRP12H48 1 μm 12 Transparent 2 x 106 pores/cm2
PTRP24H48 1 μm 24 Transparent 2 x 106 pores/cm2
PTMP06H48 5 μm 6 Transparent -
PTMP12H48 5 μm 12 Transparent -
PTMP24H48 5 μm 24 Transparent -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. PCEP06H48 Supplier MilliporeSigma Supplier No. PCEP06H48

Millicell™ hanging cell culture inserts with track-etched PET membrane promote natural growth and differentiation of various cell types

  • Individually blister-packaged inserts fit in most culture plates
  • TC-treated and gamma sterilized
  • Pore sizes range from 0.4 to 8.0 μm
  • Low pore density membranes are optically transparent
  • High pore density membranes are optically translucent
  • Applications include co-culture, migration and invasion assays, toxicity testing, transport and permeability assays and organotypic Air-Liquid Interface (ALI) culture

Specifications

Culture Area 4.5 cm2
Material PET
Volume (Metric) Well 4 mL
Volume (Metric) Working 2 mL
Pore Size 8 μm
Type Cell Culture Insert
For Use With (Equipment) Culture Plate
Diameter (Metric) Inner 24 mm
Diameter (Metric) Outer 27 mm
Height (Metric) 15.5 mm
Quantity 48
No. per Pack 48/Pk.
Color Transparent
Surface Treatment TC-treated
Sterility Sterile
No. of Wells 6
Packaging Individually Blister Packaged
Disposable Single-use
Autoclavable Not Autoclavable
Show More Show Less