missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Milliammeter, DC

Ideal option for classroom electricity demonstrations.

Supplier:  United Scientific™ MMA005

 View more versions of this product

Catalog No. S16005


Only null left
Explore available promotions

Description

Description

  • DC milliammeter mounted in a black plastic case.
  • Scale length of 75 mm, with ± 2.5% accuracy
  • Features 5-way binding post and an external zero-setting screw
  • Not protected against prolonged or significant overload
  • Stackable for convenient storage
Specifications

Specifications

Milliammeter
Classroom Demonstrations
0 to 100 mA
Analog
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Warning: Cancer and reproductive harm