missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Millex™-GV Sterile Syringe Filter Unit, PVDF, 0.22 μm

Catalog No. SLGV013SL
Change view
Click to view available options
Diameter (Metric):
0.4 cm
1.3 cm
3.3 cm
Quantity:
100/Pk.
50/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Quantity
SLGV013SL 1.3 cm 100/Pk.
SLGV033RS 3.3 cm 50/Pk.
SLGV004SL 0.4 cm 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SLGV013SL Supplier MilliporeSigma™ Supplier No. SLGV013SL

Ready-to-use, syringe-driven units designed for sterilizing and clarifying sterile solutions

Sterile Millex filter units are available in a larger 33 mm housing with either MCE, PVDF or PES membranes.

Faster flow rate:The larger filter surface area increases flow rate and throughput. It also makes it easier to filter solutions because it reduces the pressure required to empty the syringe.

Higher operating pressure:

Millex filters have a maximum housing pressure of 10.5 bar (150 psig), which means you can filter solutions faster than before.

Color-coded:

The color-coded band on the Millex housing clearly indicates which membrane is inside.

Manufactured for reliable performance:

Millex filters are manufactured in a controlled environment using an automated process. Human hands never touch the filter during assembly. A Certificate of Quality included in each box describes Millipore's quality standards in detail.

Choice of membranes:Millex filters are available with three membranes: Millipore Express™ (PES) membrane, mixed cellulose esters (MCE), and Durapore (PVDF).

  • Millex-GP filters feature Millipore Express (PES) membrane, which provides a unique combination of speed and six times less protein binding than competitive PES membranes. Millex-GP filters are recommended for sterile filtering protein solutions, tissue culture media, buffers and additives.
  • Millex filters with Durapore™ (PVDF) membrane are the lowest protein-binding syringe filters available. Use them to filter protein solutions. The 0.2 μm filters are sterilizing grade. Larger pore sizes are for clarification and prefiltration.
  • Millex-MF filters feature mixed cellulose esters (MCE) membrane, which provides reliable, general purpose filtration of water or other aqueous solutions. The 0.2 μm filters are sterilizing grade. Larger pore sizes are for clarification and prefiltration.Applications:Sterilization of Aqueous Solutions, Organic Solvents and Gases, Filtration of High Value Protein or Small Molecule Components.

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Meets USP Class VI Biological tests for plastics. All 33 mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Color Yellow
Disposable Single-use
Filter Type Membrane Filter
Material (Membrane) PVDF (Hydrophilic)
Pore Size 0.22 μm
Sterility Sterile
For Use With (Application) Ultra-low protein binding; Sterilization of tissue culture media, protein solutions or aqueous solutions
Hold up Volume (After Air Purge) <25 μL
Housing Material PVDF
Inlet Style Female Luer Lock
Outlet Style Male Luer Slip
Pressure Rating 100 psi
Quantity 100/Pk.
Sterilization Method Ethylene oxide sterilized
Temperature (English) Operating 113°F
Temperature (Metric) Operating 45°C
Temperature Range (English) 113°F
Temperature Range (Metric) 45°C
Tip Material Polyethylene, Polyester
Type Syringe Filter
Volume (Metric) 10 mL
Diameter (Metric) 1.3 cm
Show More Show Less