Learn More
MilliporeSigma™ Millex-GV Filter, 0.22 μm
Catalog No. SLGVV255F
Change view
Click to view available options
Pore Size: 0.22 μm
Catalog No.
SLGVV255F Pore Size: 0.22 μm
1 options
Catalog No. SLGVV255F Supplier MilliporeSigma™ Supplier No. SLGVV255F
Only null left
The Vented Millex™-GV filter unit SLGVV255F are syringe filters for sterilization of low volume aqueous solutions for direct patient care applications, where venting and low protein binding membrane are required or desired.
  • Tailored to a variety of applications based on membrane type, housing and validation rating
  • Medical Device-rated devices in some countries including vented devices for direct patient care
  • Specialized filter units to protect hemodialysis transducers from blood and moisture and for vacuum line protection
  • Low extractables and low absorption membranes for minimum sample losses
  • Faster flow rates and higher operating pressures for high performance needs
  • Applications: Sterilization of Aqueous Solutions, Organic Solvents and Gases, Filtration of High Value Protein or Small Molecule Components, Select Medical Device Applications (Some Including Direct Patient Care Depending on Country/Region)