missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIIP Polyclonal Antibody

Description
Antibody detects endogenous levels of total IIP45.
MIIP inhibits glioma cells invasion and down-regulates adhesion-and motility-associated genes such as NFKB2 and ICAM1. MIIP exhibits opposing effects to IGFBP2 on cell invasion.
Specifications
Specifications
| Antigen | MIIP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MIIP |
| Gene Accession No. | A2A7Y5, Q5JXC2 |
| Gene Alias | AA553001; AI853621; D4Wsu114e; IGFBP2-binding protein; IGFBP-2-Binding Protein, IIp45; Iip45; invasion inhibitory protein 45; invasion-inhibitory protein 45; migration and invasion inhibitory protein; migration and invasion-inhibitory protein; MIIP |
| Gene Symbols | MIIP |
| Show More |