missing translation for 'onlineSavingsMsg'
Learn More

Midecamycin, 900μg/mg

Catalog No. p-7057447
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66046ME 500 mg
AAJ6604603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66046ME Supplier Thermo Scientific Chemicals Supplier No. J66046ME

A broad spectrum antibiotic | CAS: 35457-80-8 | C41H67NO15 | 813.979 g/mol

A broad spectrum antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35457-80-8
Molecular Formula C41H67NO15
Molecular Weight (g/mol) 813.979
MDL Number MFCD00865000
InChI Key DMUAPQTXSSNEDD-RBATUFKESA-N
PubChem CID 124225192
IUPAC Name [(4R,5R,6S,7R,9R,10R,11E,13Z,16R)-6-[(2S,3R,4R,5S,6R)-4-(dimethylamino)-3-hydroxy-5-[(2S,4R,5S,6S)-4-hydroxy-4,6-dimethyl-5-propanoyloxyoxan-2-yl]oxy-6-methyloxan-2-yl]oxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-di
SMILES CCC(=O)OC1CC(=O)OC(CC=CC=CC(C(CC(C(C1OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)OC(=O)CC)(C)O)N(C)C)O)CC=O)C)O)C

Specifications

Type Midecamycin
Physical Form Powder
Melting Point 155°C to 156°C
Color White
Quantity 500 mg
Flash Point >110°C (230°F)
Boiling Point 874°C
Solubility Information Soluble in methanol or ethanol; Insoluble in water
Formula Weight 813.97
Chemical Name or Material Midecamycin, 900μg/mg
EINECSNumber 252-578-0
RTECSNumber OH4730600
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only