missing translation for 'onlineSavingsMsg'
Learn More

Wanzl Middle Shelf with Black Powerdercoat Finish

Middle Shelf for bulk shelving

$300.47 - $517.09

Specifications

Finish Powdercoat
For Use With (Application) Shelving
No. of Shelves 1
Shelf Type Middle Shelf
Color Black
View More Specs

Products 4
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
11222148
View Documents
Wanzl
TBCRSHT2436B
24 x 36 in.
Each for $300.47
Only null left
 
11222149
View Documents
Wanzl
TBCRSHT2448B
24 x 48 in.
Each for $399.49
Only null left
 
11222150
View Documents
Wanzl
TBCRSHT3636B
36 x 36 in.
Each for $386.00
Only null left
 
11222151
View Documents
Wanzl
TBCRSH3648B
36 x 48 in.
Each for $517.09
Only null left
 
Description

Description

  • Middle shelf
  • Replacement part for bulk shelving system
  • Black powdercoat finish
Specifications

Specifications

Powdercoat
1
Black
Shelving
Middle Shelf
Steel
Videos
SDS
Documents

Documents