missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl Middle Shelf with Black Powerdercoat Finish
Description
- Middle shelf
- Replacement part for bulk shelving system
- Black powdercoat finish
Specifications
Specifications
| Width (English) | 36 in. |
| For Use With (Application) | Shelving |
| Shelf Type | Middle Shelf |
| Finish | Powdercoat |
| Color | Black |
| Dimensions (L x W) | 24 x 36 in. |
| No. of Shelves | 1 |
| Material | Steel |