missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MICS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104374
Description
Antibody detects endogenous levels of total GHITM.
Required for the mitochondrial tubular network and cristae organization. Involved in apoptotic release of cytochrome c.Specifications
| MICS1 | |
| Polyclonal | |
| Unconjugated | |
| GHITM | |
| 1010001P14Rik; C77840; dermal papilla-derived protein 2; DERP2; DKFZp566C0746; FLJ26584; GHITM; growth hormone inducible transmembrane protein; growth hormone-inducible transmembrane protein; growth hormone-inducible transmembrane protein-like protein; HSPC282; MICS1; mitochondrial morphology and cristae structure 1; My021; PTD010; similar to Dermal papilla derived protein 2; Tmbim5; transmembrane BAX inhibitor motif containing 5; transmembrane BAX inhibitor motif-containing protein 5; UNQ244/PRO281 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27069 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H3K2 | |
| GHITM | |
| A synthesized peptide derived from human GHITM(Accession Q9H3K2), corresponding to amino acid residues F21-M71. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |