missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroSEQ™ ID Purification Combo Kit, with clean-up plates
GREENER_CHOICE

Catalog No. 4415506
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4415506 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4415506 Supplier Applied Biosystems™ Supplier No. 4415506

This MicroSEQ ID Purification Combo Kit includes reagents for post-PCR and post-sequencing steps in the MicroSEQ ID microbial identification workflow.

This MicroSEQ™ ID Purification Combo Kit includes reagents for post-PCR and post-sequencing steps in the MicroSEQ™ ID microbial identification workflow. It contains 3 tubes of ExoSAP-IT™ Reagent for PCR clean-up and two 96-well MicroSEQ™ ID Sequencing Cleanup Plates.

ExoSAP-IT™ Reagent: this reagent is used for quick post-PCR clean-up for downstream applications such as sequencing. It allows for removal of unconsumed dNTPs and primers remaining in the PCR mix following amplification.

ExoSAP-IT™ Reagent can be used downstream of the following PCR amplification kits:

  • MicroSEQ™ 500 16S rDNA PCR Kit
  • MicroSEQ™ Full Gene 16S rDNA PCR Kit
  • MicroSEQ™ D2 rDNA Fungal PCR Kit

MicroSEQ™ ID Sequencing Clean-up Plates: these plates are used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel filtration matrix used in the microplate cartridges provide a convenient and simple way to remove dye terminators, dNTPs, and salts, as well as residual primers from cycle sequencing reactions. The end products can be directly used for sequencing on any Applied Biosystems™ genetic analyzers.

MicroSEQ™ ID Sequencing Cleanup Plates can be used downstream of the following sequencing kits:

  • MicroSEQ™ 500 16S rDNA Sequencing Kit
  • MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ™ D2 rDNA Fungal Sequencing Kit

Specifications

Content And Storage • ExoSAP-IT™ Reagent for PCR clean-up, 3 tubes
• MicroSEQ™ ID Sequencing Cleanup Plates, 2 x 96-well
Format 96-well Plate
Sample Type dsDNA
Final Product Type DNA
For Use With (Application) Sequencing
No. of Reactions 100 Preps
Product Line MicroSEQ
Product Type ID Purification Combo Kit
Quantity 1 Kit
Target Sequencing Reactions
System Type MicroSEQ™ ID
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.