Learn More
Applied Biosystems™ MicroSEQ™ ID 16S rDNA Full Gene Library v2.1
Description
The MicroSEQ ID 16S rDNA Full Gene Library v2.1 is designed for installation with the MicroSEQ ID analysis software. It contains the complete 16S sequence from bacterial organisms that can be compared to an unknown bacterial sequence using MicroSEQ ID analysis software to identify bacteria. MicroSEQ ID analysis software enables you to identify and classify unidentified bacterial or fungal sequences by comparing them to a validated microbial library.
- A validated library of over 2300 microbial sequences ensures you get the answers you need
- Easy-to-use interface and optimized search algorithms enable quick results
- Phylogenetic trees provide a deeper understanding of your unknowns
- Security features support 21 CFR Part 11 compliance, enabling the software to integrate smoothly into your workflow
Specifications
Specifications
| Sequencing Type | Genome and DNA Sequencing |
| For Use With (Equipment) | SeqStudio Genetic Analyzers, SeqStudio Flex Genetic Analyzers |
| For Use With (Application) | Microbial Identification |
| License | Static |
| Operating System | Microsoft Windows 10 |
| Product Line | MicroSeq™ ID |
| Product Type | Microbial Identification Software |
| Quantity | 1 Ea. |
| Software | Sequence Analysis, Microbial Identification |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
The operating system requirements for MicroSEQ ID 16S rDNA Full Gene Library v2.1 are Windows 10, Enterprise LTSC 2018 Microsoft Corporation, 64 bit OS (Operating System), x64 Based Processor, Version-1809.
MicroSEQ ID 16S rDNA Full Gene Library v2.1 contains identical content to the previous generation library, MicroSEQ ID 16S rDNA Full Gene Library v2.0. MicroSEQ ID 16s rDNA Full Gene Library v2.1 is compatible with MicroSEQ ID Microbial Identification Software v4.0 whereas MicroSEQ ID 16s rDNA Full Gene Library v2.0 is compatible with MicroSEQ ID Microbial Identification Software v3.1.3.
MicroSEQ ID 16S rDNA Full Gene Library v2.1 is compatible with Google Chrome v108.0.5359.73.
MicroSEQ ID Microbial Identification Software v4.0 is compatible with 64-bit Windows 10 OS.
The computer requirements for MicroSEQ ID Microbial Identification Software v4.0 are:
- Intel Core i7-8700 CPU at 3.20 Ghz, with 16 GB of RAM and a minimum of 465 GB hard disk space
- WIN 10 IoT 64-bit
For Research Use Only. Not for use in diagnostic procedures.