Learn More
United Scientific™ Microscope Slide Mailer
Description
- These white polypropylene slide mailers included hinged covers and molded-in dividers to securely hold 3 x 1 in. microscope slides.
- Available in four sizes.
- These white polypropylene slide mailers included hinged covers and molded-in dividers to securely hold 3 x 1 in. microscope slides.
- Available in four sizes.
Specifications
Specifications
| Type | Slide Mailer |
| Slide Capacity | 1 |
| For Use With (Equipment) | Microscope Slides |