Learn More
United Scientific™ Microscope Slide Box, ABS
Classic design to safely store microscope slides.
Supplier: United Scientific™ 62611
Description
- Designed as a convenient solution for temporary storage and transferring of microscope slides.The bottom is lined with a cork pad, and the lid has a printed index for quick reference.
- Accommodates microscope slides in two sizes:
- 75.0 mm x 25.0 mm x (0.8 to 1.2 mm)
- 76.0 mm x 26.0 mm x (0.8 to 1.2 mm)
- Made of strong ABS material
- Snap closures and hinged-lids provide safe operation
- A water-proof index card is mounted inside the lids for simple specimen identification
- Cork sheet protects the slides
- The 100-place slide box includes a metal snap closure
Specifications
| Slide Storage Box | |
| 100 | |
| White | |
| ABS |