Learn More
Sartorius Microsart™ Media
Enhance the safety of microbiological quality control in the pharmaceutical and biotech industries for microbial limits testing according to the USP and EP
$817.33 - $817.33
Specifications
Product Type Growth Medium
Serum Level Serum-free
Sterility Sterile
Sterilization Method Gamma-irradiated
Formulation Solid Agar Medium
View More Specs
Products 2
Catalog Number Mfr. No. Without Additives Price Quantity  
Catalog Number Mfr. No. Without Additives Price Quantity  
14-556-292
View Documents Promotion Details
Sartorius
1432247ACN
No Sucrose
Pack of 100 Each for $817.33
Only null left
14-556-290
View Documents Promotion Details
Sartorius
1431347ACN
No Cholesterol
Pack of 100 Each for $817.33
Only null left
Description
Description
  • Ready-to-use agar media dish (prefilled with different agar media types) with active lid
  • Sterile packaged and ready to use together with Microsart filter units
  • Active lid enables touch-free transfer of membrane, without using any tweezers and reduces the risk of secondary contamination
  • Liftable interior lid provides easy access to select colonies after incubation for further analysis
Specifications
Specifications
Growth Medium
Sterile
Solid Agar Medium
Serum-free
Gamma-irradiated