missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ MicroGuard MP™ Microporous Lab Coats, No Pockets
Microporous lab coats deliver protection and comfort through a strong and durable fabric designed for worker comfort.
$114.23 - $114.23
Specifications
| Product Type | Controlled Environments Lab Coat |
|---|---|
| Gender | Unisex |
| Material | Microporous Composite Fabric |
| Color | White |
| Closure Type | Snaps |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
19472454
|
International Enviroguard™
8046M |
Medium |
Case of 30 Each for $114.23
|
|
|||||
|
19472455
|
International Enviroguard™
8046L |
Large |
Case of 30 Each for $114.23
|
|
|||||
|
19472456
|
International Enviroguard™
8046XL |
X-Large |
Case of 30 Each for $114.23
|
|
|||||
|
19472457
|
International Enviroguard™
80462XL |
2X-Large |
Case of 30 Each for $114.23
|
|
|||||
|
19472458
|
International Enviroguard™
80463XL |
3X-Large |
Case of 30 Each for $114.23
|
|
|||||
|
19472459
|
International Enviroguard™
80464XL |
4X-Large |
Case of 30 Each for $114.23
|
|
|||||
Description
- Microporous, disposable lab coats offer the balance of safety and comfort
- Fabric keeps dust and liquids out while allowing air to flow freely to keep worker cool
- Ideal for non-hazardous work environments where reliable liquid and particulate protection is needed
- Produced with state-of-the-art lamination technology to combine a soft non-woven fabric with a microporous film that repels liquids
- Breathable fabric lets moisture vapor escape
- No pockets
- Folded collar
- Environmentally friendly- can be recycled after use
- Elastic wrists offer a snug, secure fit to keep sleeves in place to prevent skin exposure
Specifications
| Controlled Environments Lab Coat | |
| Microporous Composite Fabric | |
| Snaps | |
| Single-use | |
| None |
| Unisex | |
| White | |
| Elastic | |
| Non-sterile | |
| Serged |