missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ MicroGuard MP™ Microporous Lab Coats, No Pockets
Microporous lab coats deliver protection and comfort through a strong and durable fabric designed for worker comfort.
Supplier: International Enviroguard™ 8046L
Description
- Microporous, disposable lab coats offer the balance of safety and comfort
- Fabric keeps dust and liquids out while allowing air to flow freely to keep worker cool
- Ideal for non-hazardous work environments where reliable liquid and particulate protection is needed
- Produced with state-of-the-art lamination technology to combine a soft non-woven fabric with a microporous film that repels liquids
- Breathable fabric lets moisture vapor escape
- No pockets
- Folded collar
- Environmentally friendly- can be recycled after use
- Elastic wrists offer a snug, secure fit to keep sleeves in place to prevent skin exposure
Specifications
| Controlled Environments Lab Coat | |
| Unisex | |
| White | |
| Elastic | |
| Non-sterile | |
| Serged |
| Large | |
| Microporous Composite Fabric | |
| Snaps | |
| Single-use | |
| None |