Learn More
MicroGuard MP™ Microporous Coveralls, Elastic Wrists and Ankles
Microporous coveralls deliver protection and comfort through a strong and durable fabric.
$167.23 - $198.60
Specifications
Product Type General Purpose Coveralls
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19472763
View Documents
MicroGuard MP™
8013M
Medium N/A
Only null left
19472762
View Documents
MicroGuard MP™
8013L
Large N/A
Only null left
19472764
View Documents
MicroGuard MP™
8013XL
X-Large N/A
Only null left
19472758
View Documents
MicroGuard MP™
80132XL
2X-Large N/A
Only null left
19472759
View Documents
MicroGuard MP™
80133XL
3X-Large N/A
Only null left
19472760
View Documents
MicroGuard MP™
80134XL
4X-Large N/A
Only null left
19472761
View Documents
MicroGuard MP™
80135XL
5X-Large N/A
Only null left
Description
Description
  • Comfortable state-of-the-art lamination technology allows for maximum breathability.
  • Soft non-woven fabric combined with a microporous film allows moisture vapor to escape, so heat and moisture are not trapped near the body.
  • Elastic wrists and elastic ankles.
  • Coveralls offer base-level protection against light liquid splashes and nonhazardous particulates providing the optimal balance of comfort, durability, and protection.
  • Roomy, seamless shoulder area resists tears and provides more protection in key exposure areas.
  • Wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric.
  • Elastic back stretches with user for more comfort and better tear resistance.
  • Elastic wrists and ankles offer a secure fit to help prevent skin exposure.
  • Environmentally friendly, can be recycled after use.
  • Attached hood provides additional protection of the neck and shoulders.
Specifications
Specifications
General Purpose Coveralls
Microporous Fabric
Single-use
Elastic
Collared
Front Zipper with Storm Flap
Non-sterile
Abrasion, Caustics, Liquid, Particle, Water
8013
Unisex
White
Elastic
Elastic
Unhooded
Serged
General
Elastic
None
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'