missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MicroGuard MP™ Microporous Coveralls, Elastic Wrists and Ankles
Microporous coveralls deliver protection and comfort through a strong and durable fabric.
Supplier: MicroGuard MP™ 80133XL
Description
- Comfortable state-of-the-art lamination technology allows for maximum breathability.
- Soft non-woven fabric combined with a microporous film allows moisture vapor to escape, so heat and moisture are not trapped near the body.
- Elastic wrists and elastic ankles.
- Coveralls offer base-level protection against light liquid splashes and nonhazardous particulates providing the optimal balance of comfort, durability, and protection.
- Roomy, seamless shoulder area resists tears and provides more protection in key exposure areas.
- Wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric.
- Elastic back stretches with user for more comfort and better tear resistance.
- Elastic wrists and ankles offer a secure fit to help prevent skin exposure.
- Environmentally friendly, can be recycled after use.
- Attached hood provides additional protection of the neck and shoulders.
Specifications
| General Purpose Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Elastic | |
| Unhooded | |
| Serged | |
| General | |
| Elastic | |
| None |
| 3X-Large | |
| Microporous Fabric | |
| Single-use | |
| Elastic | |
| Collared | |
| Front Zipper with Storm Flap | |
| Non-sterile | |
| Abrasion, Caustics, Liquid, Particle, Water | |
| 8013 |