missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microplate Shakers with 4-Place Platform

For life science, immunoassay. shaking ELISA plates, staining cells for flow cytometry,shaking plates in cold room, food and beverage, histamine in cheese, vitamins in milk testing applications

$1,259.30

Specifications

Orbit 2.5 mm
Load Bearing Capacity (Metric) 5 kg
Speed Control Accuracy Speed Accuracy: ±1 rpm
Speed Range 150 to 1000 rpm
Voltage 100/240 V
View More Specs

Products 1
Catalog Number Mfr. No. Plug Type Price Quantity & Availability  
Catalog Number Mfr. No. Plug Type Price Quantity & Availability  
88861023
View Documents Promotion Details
Fisherbrand™
88861023
US Plug
Each for $1,259.30
Only null left
 
Description

Description

  • Variable shaking speed: 150 to 1000rpm 
  • Quick and easy fitting or any standard-depth microplates 
  • Continuous or timed operation, with automatic switch off 
  • Display will show last used settings, even after power has been turned off.
  • Continue to run by last set speed and the remaining time when the power recovered.
  • Operating temperature 5°C to 40°C allows for use in refrigerators, cold rooms and non-CO2 incubators
  • Holds up to four microplates  
  • Easy-to-use electronic timer ensures accurate count-down and repeatability of time sensitive incubations
Specifications

Specifications

2.5 mm
Speed Accuracy: ±1 rpm
100/240 V
Aluminum Platform
415 x 293 x 143mm
0 min. to 99:59 hr.
5 kg
150 to 1000 rpm
5°C to 40°C
320 x 260 mm
LED
Videos
SDS
Documents

Documents