missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microplate Shakers with 4-Place Platform

Catalog No. 88861023
Change view
Click to view available options
Plug Type:
US Plug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type
88861023 US Plug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88861023 Supplier Fisherbrand™ Supplier No. 88861023

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

4-place microplate clamps on aluminum platform

For life science, immunoassay. shaking ELISA plates, staining cells for flow cytometry,shaking plates in cold room, food and beverage, histamine in cheese, vitamins in milk testing applications

  • Variable shaking speed: 150 to 1000rpm
  • Quick and easy fitting or any standard-depth microplates
  • Continuous or timed operation, with automatic switch off
  • Display will show last used settings, even after power has been turned off.
  • Continue to run by last set speed and the remaining time when the power recovered.
  • Operating temperature 5°C to 40°C allows for use in refrigerators, cold rooms and non-CO2 incubators
  • Holds up to four microplates
  • Easy-to-use electronic timer ensures accurate count-down and repeatability of time sensitive incubations

Specifications

Orbit 2.5 mm
Load Bearing Capacity (Metric) 5 kg
Speed Control Accuracy Speed Accuracy: ±1 rpm
Speed Range 150 to 1000 rpm
Voltage 100/240 V
Temperature Range (Metric) 5°C to 40°C
Product Type Microplate Shaker
Plug Type US Plug
Material Aluminum Platform
Dimensions (D x W x H) Interior 297 x 156 x12 mm
Dimensions (LxW) Platform 320 x 260 mm
Dimensions (L x W x H) 415 x 293 x 143mm
Model Fisherbrand Microplate Shaker with 4-Place Platform
Display LED
Includes 4-place microplate clamps on aluminum platform
Timer Range 0 min. to 99:59 hr.
Show More Show Less