Learn More
Gilson™ MICROMAN™ E Positive Displacement Pipette PROMO
The ergonomically designed MICROMAN™ E Positive Displacement Pipette, along with disposable capillary piston tips, provide the highest precision when pipetting viscous, non-aqueous liquids while protecting the pipette from contamination.
Supplier:  Gilson™ FD10004GPM
 View more versions of this product
Catalog No. FD10004GPM
Only null left
Explore available promotions
Description
Description
MICROMAN™ E positive displacement pipettes with capillary pistons offer highest precision when pipetting problem liquids such as oils, cosmetics, blood, glycerol, radioactive samples, RNA, DNA, and more.
  • Ergonomic design to increase pipetting performance.
  • Patented QuickSnap feature for Capillary Piston fitting System.
  • Volume securing button to reduce risk of accidental volume change during pipetting cycles.
  • Comfortable and easy to use with new handle shape and added finger hook.
  • Fully visible volumeter.
  • Large pushbutton for allows easy aspirate and dispense.
  • For complete and rapid pipetting of viscous, volatile, dense, hot or cold liquids.
  • Avoids leaks when pipetting volatile (high vapor pressure) liquids.
  • Avoid cross-contamination or residual sample carry-over.
  • For tips, see F148412G or F148413G.
Specifications
Specifications
CP Tips
Yellow
M100E
Positive Displacement Pipette
For pipetting viscous, volatile, and problem liquids
12.6 in.
1.38 in.
Autoclavable
Polypropylene
1
10 to 100 μL
1 Ea.
4.33 in.
Certified Quality
Product Suggestions
Product Suggestions