missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ MICROMAN™ E Positive Displacement Pipette PROMO

Catalog No. FD10005GPM
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
10 to 100 μL
20 to 50 μL
3 to 25 μL
50 to 250 μL
Model:
M100E
M10E
M250E
M25E
M50E
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model
FD10005GPM 50 to 250 μL M250E
FD10001GPM 1 to 10 μL M10E
FD10002GPM 3 to 25 μL M25E
FD10003GPM 20 to 50 μL M50E
FD10004GPM 10 to 100 μL M100E
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. FD10005GPM Supplier Gilson™ Supplier No. FD10005GPM
Get Up to 50% Off Liquid Handling Solutions
View all promotions

The ergonomically designed MICROMAN™ E Positive Displacement Pipette, along with disposable capillary piston tips, provide the highest precision when pipetting viscous, non-aqueous liquids while protecting the pipette from contamination.

MICROMAN™ E positive displacement pipettes with capillary pistons offer highest precision when pipetting problem liquids such as oils, cosmetics, blood, glycerol, radioactive samples, RNA, DNA, and more.

  • Ergonomic design to increase pipetting performance.
  • Patented QuickSnap feature for Capillary Piston fitting System.
  • Volume securing button to reduce risk of accidental volume change during pipetting cycles.
  • Comfortable and easy to use with new handle shape and added finger hook.
  • Fully visible volumeter.
  • Large pushbutton for allows easy aspirate and dispense.
  • For complete and rapid pipetting of viscous, volatile, dense, hot or cold liquids.
  • Avoids leaks when pipetting volatile (high vapor pressure) liquids.
  • Avoid cross-contamination or residual sample carry-over.
  • For tips, see F148412G or F148413G.

Specifications

Compatible Tips CP Tips
Autoclavable Autoclavable
Color Red
Material Polypropylene
Model M250E
Number of Channels 1
Type Positive Displacement Pipette
Volume (Metric) 50 to 250 μL
For Use With (Application) For pipetting viscous, volatile, and problem liquids
Quantity 1 Ea.
Length (English) 4.25 in.
Width (English) 12.48 in.
Height (English) 1.42 in.
Fit for Purpose Certified Quality
Show More Show Less