missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter Limited Volume Inserts for 9mm and 11mm Autosampler Vials, Silanized

Catalog No. 15111472
Change view
Click to view available options
Volume (Metric) Insert:
300 μL
350 μL
Base Style:
Conical with Pulled Point
Conical with Spring
Flat Bottom
Size (Metric) Opening:
9 mm
11 mm
Quantity:
100 Pk.
1000 Pk.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Insert Base Style Size (Metric) Opening Quantity
15111472 300 μL Conical with Spring 11 mm 100 Pk.
15111480 350 μL Flat Bottom 11 mm 100 Pk.
15111476 350 μL Conical with Pulled Point 11 mm 100 Pk.
15111006 350 μL Conical with Pulled Point 9 mm 1000 Pk.
15111005 350 μL Conical with Pulled Point 9 mm 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 15111472 Supplier DWK Life Sciences Supplier No. 110000100S

MicroLiter limited volume inserts are made from Type 1 borosilicate glass or polypropylene and meet strict tolerances. Conical inserts, inserts with plastic springs, and flat bottom inserts are available to assist in consistent sampling of low-volume samples.

Specifications

Color Clear
Material Borosilicate Glass (Type 1)
Size (Metric) Opening 11 mm
Volume (Metric) Insert 300 μL
Base Style Conical with Spring
Quantity 100 Pk.
For Use With (Equipment) 11 mm Autosampler Vials - MicroLiter