Learn More
DWK Life Sciences MicroLiter Glass Limited Volume Inserts for 9 mm Autosampler Vials, Untreated
MicroLiter limited volume inserts are made from Type 1 borosilicate glass or polypropylene and meet strict tolerances. Conical inserts, inserts with plastic springs, and flat bottom inserts are available to assist in consistent sampling of low-volume samples.
$129.86 - $1,272.69
Specifications
| Color | Clear |
|---|---|
| Material | Borosilicate Glass (Type 1) |
| Volume (Metric) Insert | 350 μL |
| Base Style | Conical with Pulled Point |
| Product Line | MicroLiter |
Specifications
| Clear | |
| 350 μL | |
| MicroLiter |
| Borosilicate Glass (Type 1) | |
| Conical with Pulled Point | |
| 9 mm Autosampler Vials - MicroLiter |