missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial Septa

Catalog No. 15111049
Change view
Click to view available options
Color:
Orange
Red
Red, White
White
White, Blue
Septum:
ETFE/Silicone
FEP/Natural Rubber
PTFE
PTFE/Silicone
PTFE/Silicone/PTFE
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum
15111049 Orange ETFE/Silicone
15111014 White PTFE
15111015 Orange FEP/Natural Rubber
15111065 Red, White PTFE/Silicone
15111085 White, Blue PTFE/Silicone
15111103 Red PTFE/Silicone/PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 15111049 Supplier DWK Life Sciences Supplier No. 090022

MicroLiter septa for 9mm Screwthread Autosampler Vials are constructed with a wide array of chemical compatibilities to suit a variety of chromatography applications.

Specifications

Color Orange
Pre-slit No
Septum ETFE/Silicone
Quantity 100
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter