missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial Caps with PTFE/Silicone/PTFE Septa

Catalog No. 15111121
Change view
Click to view available options
Color:
Black
Blue
Green
Natural
Red
Yellow
Product Type:
Crimp Seal
Screw Cap
Quantity:
100
2000
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Quantity
15111121 Yellow Crimp Seal 100
15111104 Blue Screw Cap 100
15111105 Blue Screw Cap 2000
15111107 Green Screw Cap 100
15111108 Green Screw Cap 2000
15111110 Black Screw Cap 100
15111111 Black Screw Cap 2000
15111113 Natural Screw Cap 100
15111114 Natural Screw Cap 2000
15111115 Natural Crimp Seal 100
15111116 Red Screw Cap 100
15111117 Red Screw Cap 2000
15111120 Yellow Screw Cap 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 15111121 Supplier DWK Life Sciences Supplier No. 090040YT

Available in multiple colors and package sizes.

Specifications

Material (Liner) PTFE/Silicone/PTFE
Color Yellow
Material Polypropylene
Pre-slit No
Septum PTFE/Silicone/PTFE
Quantity 100
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Type Crimp Seal