missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial Caps with FKM Fluoroelastomer Septa

Catalog No. 15111123
Change view
Click to view available options
Color:
Green
Natural
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15111123 Green
15111125 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15111123 Supplier DWK Life Sciences Supplier No. 090080G
Only null left

Available in multiple colors and package sizes.

Specifications

Material (Liner) FKM
Color Green
Material Polypropylene
Pre-slit No
Septum FKM
Quantity 100
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Type Screw Cap