Learn More
DWK Life Sciences MicroLiter 9 mm Screw Thread Autosampler Vial Caps with FEP/Natural Rubber Septa
Available in multiple colors and package sizes.
Supplier: DWK Life Sciences 090020G
Chemical Identifiers
| Polypropylene | |
| FEP, Natural Rubber, FEP | |
| 9 mm | |
| Screw Cap |
| No | |
| MicroLiter | |
| 9 mm Autosampler Vials - MicroLiter |
Specifications
| Green | |
| No | |
| 100 | |
| 9 mm | |
| Screw Cap |
| Polypropylene | |
| FEP, Natural Rubber, FEP | |
| MicroLiter | |
| 9 mm Autosampler Vials - MicroLiter |