Learn More
DWK Life Sciences MicroLiter 9 mm Clear Glass Screw Thread Autosampler Vial, Cap, and Septa Assembled Kits with PTFE/Silicone Septa
Save time and money with vial, cap, and septa kits that arrive conveniently packaged and preassembled.
Supplier:  DWK Life Sciences 091030GA
 View more versions of this product
Catalog No. 15111187
Only null left
Description
Description
Available in multiple colors and septa formats and with or without writing patch.
Specifications
Chemical Identifiers
Clear
Borosilicate Glass (Type 1)
100
Polypropylene
9 mm
12 mm
2 mL
MicroLiter
Screw Cap
32 mm
Specifications
Green
12 mm
No
PTFE, Silicone
MicroLiter
Screw Cap
No
Clear
Borosilicate Glass (Type 1)
2 mL
100
Polypropylene
9 mm
32 mm
Product Suggestions
Product Suggestions