missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 9 mm ScrewTin Autosampler Vial Caps with PTFE/Silicone Septa

Catalog No. 15111074
Change view
Click to view available options
Color:
Black
Blue
Green
Natural
Yellow
Pre-slit:
No
Yes
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Pre-slit
15111074 Black No
15111068 Blue No
15111088 Blue Yes
15111071 Green No
15111077 Natural No
15111095 Natural Yes
15111083 Yellow No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 15111074 Supplier DWK Life Sciences Supplier No. 090030KT

Available in multiple colors and package sizes.

Specifications

Color Black
Material Polypropylene
Pre-slit No
Septum PTFE, Silicone
No. per Pack 100
Product Line MicroLiter
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Type Screw Cap