missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 9 mm ScrewTin Autosampler Vial Caps with FEP/Natural Rubber Septa

Catalog No. 15111018
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15111018 Blue
15111021 Green
15111030 Red
15111033 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15111018 Supplier DWK Life Sciences Supplier No. 090020BT

Available in multiple colors and package sizes.

Specifications

Color Blue
Material Polypropylene
Pre-slit No
Septum FEP, Natural Rubber, FEP
No. per Pack 100
Product Line MicroLiter
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Type Screw Cap