missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 9 mm ILT LC-Clean™ Screw Thread Autosampler Vial Caps with PTFE/Silicone Septa

Catalog No. 15111102
Change view
Click to view available options
Color:
Blue
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15111102 Gray
15111101 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15111102 Supplier DWK Life Sciences Supplier No. 090037GR

Available in multiple colors and package sizes.

Specifications

Color Gray
Material Polypropylene
Pre-slit No
Septum PTFE, Silicone
No. per Pack 100
Product Line MicroLiter
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Type Screw Cap