missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 20mm Magnetic Crimp Top Headspace Vial Seals

Catalog No. 15111716
Change view
Click to view available options
Color:
Gold
Material (Liner):
Natural PTFE/Blue Silicone
Tan PTFE/White silicone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material (Liner)
15111716 Gold Natural PTFE/Blue Silicone
15111735 Gold Tan PTFE/White silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15111716 Supplier DWK Life Sciences Supplier No. 200050MS

MicroLiter 20mm crimp top seals are available in multiple colors and septa types.

Magnetic crimp seal

Specifications

Material (Liner) Natural PTFE/Blue Silicone
Color Gold
Material Steel
Pre-slit No
Septum PTFE/Silicone
Quantity 100
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Product Type Crimp Cap