Learn More
DWK Life Sciences MicroLiter 20 mm Crimp Top Headspace Vial Septas and Stoppers
Butyl stoppers can be used when PTFE is not required. Butyl/PTFE septa are an economical choice with low background peaks.
Supplier:  DWK Life Sciences 200025
 View more versions of this product
Catalog No. 15111670
Only null left
Description
Description
Molded PTFE/butyl septa offer the tightest seal of all PTFE items, as well as low background peaks. PTFE/silicone septa are excellent for volatiles. Aluminum/silicone septa are also excellent for volatiles, but not suitable for repeat injections.
Specifications
Chemical Identifiers
No
20 mm
20 mm Headspace Seals - MicroLiter
Specifications
Gray
No
20 mm
Stopper
Butyl
100
20 mm Headspace Seals - MicroLiter
Product Suggestions
Product Suggestions