missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 20 mm Crimp Top Headspace Vial Septas and Stoppers

Catalog No. 15111657
Change view
Click to view available options
Color:
Black
Gray
White, Tan
Material:
Butyl
PTFE, Silicone
Product Type:
3-leg Stopper
Septa
Stopper
No. per Pack:
100
1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material Product Type No. per Pack
15111657 Black Butyl Septa 100
15111722 White, Tan PTFE, Silicone Septa 1000
15111670 Gray Butyl Stopper 100
15111671 Gray Butyl Stopper 1000
15111672 Gray Butyl 3-leg Stopper 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 15111657 Supplier DWK Life Sciences Supplier No. 200021

Butyl stoppers can be used when PTFE is not required. Butyl/PTFE septa are an economical choice with low background peaks.

Molded PTFE/butyl septa offer the tightest seal of all PTFE items, as well as low background peaks. PTFE/silicone septa are excellent for volatiles. Aluminum/silicone septa are also excellent for volatiles, but not suitable for repeat injections.

Specifications

Color Black
Material Butyl
Pre-slit No
No. per Pack 100
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Seals - MicroLiter
Product Type Septa