missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 20 mm AlumiTin Crimp Top Headspace Vial Seals

Catalog No. 15111719
Change view
Click to view available options
Color:
Blue
Red
Silver
Septum:
Natural PTFE, Blue Silicone
PTFE, Butyl
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum
15111719 Red Natural PTFE, Blue Silicone
15111688 Silver PTFE, Butyl
15111709 Blue Natural PTFE, Blue Silicone
15111717 Silver Natural PTFE, Blue Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15111719 Supplier DWK Life Sciences Supplier No. 200050RT
Only null left

MicroLiter 20 mm crimp top seals are available in multiple colors and septa types.

Specifications

Color Red
Material Aluminum
Pre-slit No
Septum Natural PTFE, Blue Silicone
No. per Pack 100
Product Line MicroLiter
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Product Type Crimp Cap