missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 20 mm Aluminum Crimp Top Headspace Vial Seals

Catalog No. 15111673
Change view
Click to view available options
Septum:
Aluminum, Silicone
Butyl Stopper
Chemically Inert Butyl
Natural PTFE, Blue Silicone
PTFE, Butyl
PTFE, Molded Butyl
Tan PTFE, White Silicone
No. per Pack:
100
1000
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum No. per Pack
15111673 Butyl Stopper 100
15111658 Chemically Inert Butyl 100
15111678 PTFE, Butyl 100
15111691 PTFE, Molded Butyl 100
15111703 Natural PTFE, Blue Silicone 100
15111723 Tan PTFE, White Silicone 100
15111724 Tan PTFE, White Silicone 1000
15111740 Aluminum, Silicone 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 15111673 Supplier DWK Life Sciences Supplier No. 200025AS

MicroLiter 20 mm crimp top seals are available in multiple colors and septa types.

Specifications

Color Silver
Material Aluminum
Pre-slit No
Septum Butyl Stopper
No. per Pack 100
Product Line MicroLiter
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Product Type Crimp Cap