missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 20 mm Large Opening Extended Apron Magnetic Crimp Top Headspace Vial Seals

Catalog No. 15111636
Change view
Click to view available options
Color:
Gold
Silver
No. per Pack:
100
1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Pack
15111636 Silver 1000
15111638 Gold 1000
15111637 Gold 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15111636 Supplier DWK Life Sciences Supplier No. 200000MLB

MicroLiter 20 mm large opening crimp top seals are available in multiple colors and septa types.

Magnetic crimp seal

Specifications

Color Silver
Material Steel
Septum None
No. per Pack 1000
Product Line MicroLiter
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Septa - MicroLiter
Product Type Crimp Cap