missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ MicroLiter™ 11 mm Crimp Top Autosampler Vial Seals

Catalog No. 15111518
Change view
Click to view available options
Color:
Blue
Gold
Green
Red
Silver
Septum:
ETFE, Silicone
FEP, Natural Rubber
PTFE, Silicone
PTFE, Silicone, PTFE
Quantity:
100 Caps/Pk.
2000 Caps/Pk.
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum Quantity
15111518 Silver PTFE, Silicone 2000 Caps/Pk.
15111524 Silver PTFE, Silicone, PTFE 2000 Caps/Pk.
15111483 Silver FEP, Natural Rubber 100 Caps/Pk.
15111484 Silver FEP, Natural Rubber 2000 Caps/Pk.
15111506 Silver ETFE, Silicone 100 Caps/Pk.
15111517 Silver PTFE, Silicone 100 Caps/Pk.
15111523 Silver PTFE, Silicone, PTFE 100 Caps/Pk.
15111509 Blue ETFE, Silicone 100 Caps/Pk.
15111491 Red FEP, Natural Rubber 100 Caps/Pk.
15111513 Red ETFE, Silicone 100 Caps/Pk.
15111511 Green ETFE, Silicone 100 Caps/Pk.
01549776 Green PTFE, Silicone 100 Caps/Pk.
15111522 Gold PTFE, Silicone 100 Caps/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 15111518 Supplier DWK Life Sciences™ Supplier No. 110030AB
Only null left

MicroLiter™'s 11 mm, crimp top autosampler vial seals are available in multiple colors and septa types.

  • Crimp top vials specific to the MicroLiter™ autosampler

Specifications

Color Silver
Material Aluminum
Pre-slit No
Septum PTFE, Silicone
Product Line MicroLiter
Quantity 2000 Caps/Pk.
Closure Size 11 mm
For Use With (Equipment) Autosampler Vials
Product Type Crimp Cap