Learn More
MilliporeSigma Microfil™ Funnel with S-Pak™, White
White mixed cellulose esters gridded filter with funnel.
Supplier: MilliporeSigma MIHVWP100
Description
Microfil funnels and filters assuring leak-free operation and uniform microorganism recoveries as well as an easy transfer of the membrane with forceps and prevents accidentally touching the filtration area.
The Microfil filtration system is a simple and reliable solution for routine microbiological testing. These ready-to-use 100 or 250 mL Microfil funnels, white gridded S pak filter system eliminate the need for washing and sterilizing after each test thus making the equipment handling easier and more efficient for general microbiological analysis. The push-fit design of the Microfil funnels seals tightly to manifold supports without the need for clamps or O-rings. Unlike with conventional filter holders, incomplete sealing at the membrane/funnel interface is eliminated resulting in uniform recovery of organisms.
Spécifications
| EZ-Fit Filtration Manifold | |
| 0.45 μm | |
| Sterile | |
| Funnel and Filter |
| MCE | |
| 10 to 100 mL | |
| White |