missing translation for 'onlineSavingsMsg'
Learn More

Microcosmic Salt Solution, LabChem™

Catalog No. LC169811
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC169811 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC169811 Supplier LabChem Supplier No. LC169811

Specifications

Chemical Name or Material Microcosmic Salt Solution
Color Colorless
CAS 7783-13-3, 7732-18-5
CAS Max % 95, 5
Linear Formula H13NNaO8P
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 209.07
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Gaseous Ammonia; Phosphorus oxides
Quantity 500 mL
Physical Form Liquid
Show More Show Less