missing translation for 'onlineSavingsMsg'
Learn More

Micro Particles Based on Silicon Dioxide, Size: 1.0 m, MilliporeSigma™ Supelco™

$415.86

Specifications

Concentration 5% (solids)
Content And Storage 2°C to 8°C
Synonym Beads based on silicon dioxide, microsize; Silicon dioxide beads
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
111004308
View Documents Promotion Details
N/A 5 mL
Each for $415.86
Only null left
 
Description

Description

Specifications

Specifications

5% (solids)
2°C to 8°C
Beads based on silicon dioxide, microsize; Silicon dioxide beads
Videos
SDS
Documents

Documents