missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micro Particles Based on Silicon Dioxide, Size: 1.0 m, MilliporeSigma™ Supelco™
Specifications
Specifications
| Concentration | 5% (solids) |
| Content And Storage | 2°C to 8°C |
| Quantity | 5 mL |
| Synonym | Beads based on silicon dioxide, microsize; Silicon dioxide beads |